missing translation for 'onlineSavingsMsg'
Learn More

Samotolisib, MedChemExpress

Product Code. 16475228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475228

Brand: Medchem Express HY12513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Samotolisib (LY3023414) potently and selectively inhibits class I PI3K isoforms, DNA-PK, and mTORC1/2 with IC50s of 6.07 nM, 77.6 nM, 38 nM, 23.8 nM, 4.24 nM and 165 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ, DNA-PK and mTOR, respectively. Samotolisib potently inhibits mTORC1/2 at low nanomolar concentrations.

Samotolisib (LY3023414) potently and selectively inhibits class I PI3K isoforms, DNA-PK, and mTORC1/2 with IC50s of 6.07 nM, 77.6 nM, 38 nM, 23.8 nM, 4.24 nM and 165 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ, DNA-PK and mTOR, respectively. Samotolisib potently inhibits mTORC1/2 at low nanomolar concentrations.

Specifications

Chemical Name or Material Samotolisib
CAS 1386874-06-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H26N4O3
Quantity 10 mg
Synonym LY3023414
Solubility Information DMSO : 50 mg/mL (123.01 mM; Need ultrasonic)
SMILES O=C(N1C[C@@H](OC)C)N(C)C2=C1C3=CC(C4=CC(C(C)(O)C)=CN=C4)=CC=C3N=C2
Molecular Weight (g/mol) 406.48
Formula Weight 406.48
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.