Learn More
Thermo Scientific™ Samco™ Transfer Pipettes
GREENER_CHOICE
Product Code. p-4525493
Click to view available options
Volume (Metric):
0.6 mL
1.05 mL
1.8 mL
3.1 mL
4.9 mL
5.5 mL
7.5 mL
7.7 mL
8.0 mL
8.6 mL
9.5 mL
Sterility:
Non-sterile
Nonsterile
Sterile
Bulb Draw (Metric):
0.3 mL
0.9 mL
1.05 mL
1.8 mL
2.2 mL
2.4 mL
2.8 mL
3.2 mL
3.5 mL
4.6 mL
6.0 mL
6.2 mL
Packaging Type:
20/Bag, 25 Bags/Pk, 8 Pks/Cs.
20/Bag, 400/Pk
20/Bag, 500/Pk
4000/Cs
Bulk 250/Pk
Bulk 400/Pk
Bulk 500/Pack
Bulk 500/Pk
Bulk, 400/Pk.
Bulk, 500/pk
Individually Wrapped 250/Pk
Individually Wrapped 400/Pk
Individually Wrapped 500/Pk
Individually Wrapped, 400/Pk
Individually Wrapped, 500/pk
Unit Size:
Pack of 250
Pack of 400
Pack of 4000
Pack of 500
Product Code. 15307813
Brand: Thermo Scientific™ 202
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for: Research Applications
TRUSTED_SUSTAINABILITY
Specifications
Bulb Draw (Metric) 3.5 mL
Description General Purpose Transfer Pipettes, Standard Bulb, 7.7mL
Diameter (Metric) Outer 7.9 mm
Material LDPE
Sterility Non-sterile
Drops/mL 21
Graduated No
Packaging Type Bulk 500/Pk
Volume (Metric) 7.7 mL
Length (Metric) 15.6 cm
Type General Purpose Transfer Pipet