missing translation for 'onlineSavingsMsg'
Learn More

Salinomycin sodium salt, MP Biomedicals™

Product Code. 11444341 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 11444341

Brand: MP Biomedicals 0215204350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55721-31-8
Molecular Formula C42H69NaO11
Molecular Weight (g/mol) 772.993
MDL Number MFCD00167109
InChI Key YPZYGIQXBGHDBH-IETZRCTKSA-M
Synonym Coxistal
PubChem CID 124080880
IUPAC Name sodium;(2S)-2-[(2R,5S,6S)-6-[(2R,3R,4R,6S)-6-[(3R,5S,7R,9S,10S,12R,15R)-3-[(2S,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.5^{7}.3^{5}]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methylox
SMILES CCC(C1CCC(C(O1)C(C)C(C(C)C(=O)C(CC)C2C(CC(C3(O2)C=CC(C4(O3)CCC(O4)(C)C5CCC(C(O5)C)(CC)O)O)C)C)O)C)C(=O)[O-].[Na+]

Specifications

Type Salinomycin Sodium Salt
Physical Form Powder
Color White
Quantity 50 mg
Formula Weight 772.98
Chemical Name or Material Salinomycin sodium salt, Molecular biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.