missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salinomycin sodium salt, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
Specifications
Specifications
| Type | Salinomycin Sodium Salt |
| Physical Form | Powder |
| Color | White |
| Quantity | 5 mg |
| Formula Weight | 772.98 |
| Chemical Name or Material | Salinomycin sodium salt, Molecular biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction