Learn More
Salinomycin sodium salt, MedChemExpress
Product Code. 16413367
Encompass_Preferred
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Product Code. 16413367
Brand: Medchem Express HY17439
MedChemExpress Salinomycin sodium salt (Salinomycin sodium), an antibiotic potassium ionophore, is a potent inhibitor of Wnt/β-catenin signaling. Salinomycin sodium salt (Salinomycin sodium) acts on the Wnt/Fzd/LRP complex, blocks Wnt-induced LRP6 phosphorylation, and causes degradation of the LRP6 protein. Salinomycin sodium salt (Salinomycin sodium) shows selective activity against human cancer stem cells.
Salinomycin sodium salt (Salinomycin sodium), an antibiotic potassium ionophore, is a potent inhibitor of Wnt/β-catenin signaling. Salinomycin sodium salt (Salinomycin sodium) acts on the Wnt/Fzd/LRP complex, blocks Wnt-induced LRP6 phosphorylation, and causes degradation of the LRP6 protein. Salinomycin sodium salt (Salinomycin sodium) shows selective activity against human cancer stem cells.
Specifications
Chemical Name or Material Salinomycin sodium salt
Physical Form Solid
CAS 55721-31-8
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C42H69NaO11
For Use With (Application) Cancer-Kinase/protease
Synonym Salinomycin sodium Sodium salinomycin
Solubility Information DMSO : 100 mg/mL (129.37 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H][C@]1([C@](C)(CC2)O[C@]32[C@H](O)C=C[C@]4(O[C@]([H])([C@@H](CC)C([C@@H](C)[C@@H](O)[C@H](C)[C@]5([H])O[C@]([C@@H](CC)C(O[Na])=O)([H])CC[C@@H]5C)=O)[C@@H](C)C[C@H]4C)O3)CC[C@@](CC)(O)[C@H](C)O1
Molecular Weight (g/mol) 772.98
Formula Weight 772.98
Grade Research