missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Salinomycin ReadyMade™ Solution

Product Code. 30044050
Encompass_Preferred
Click to view available options
Quantity:
0.5 mL
10 x 0.5 mL
Unit Size:
0.5mL
5mL
This item is not returnable. View return policy

Product Code. 30044050

Brand: TOKU-E™ S13810X0.5ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Salinomycin ReadyMade™ Solution is provided as a sterile-filtered solution of Salinomycin Sodium in water.

  • CAS Number: 55721-31-8
  • Molecular Formula: C42H69NaO11
  • Molecular Weight: 772.98 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Solution
  • Source: Streptomyces albus
  • Spectrum: Salinomycin targets primarily the Gram-positive cell wall to allow ion transport into the cell. Gram-negative organisms are unaffected by Salinomycin because of their additional outer membrane. Salinomycin is also effective against mycobacteria, some filamentous fungi, and Coccidia
  • Mechanism of Action: Salinomycin interacts with the Gram-positive cell membrane which decreases control of ion permeability. This allows ions (K+, Na+, Ca+, Mg2+) to accumulate inside the cell to toxic levels

Specifications

Chemical Name or Material Salinomycin Monosodium Salt
Physical Form Liquid
CAS 55721-31-8
Source Streptomyces albus
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Colorless
Concentration 2 mg/mL
Quantity 10 x 0.5 mL
Purity Grade Notes >90% (HPLC)
Molecular Formula C42H69NaO11
Product Line ReadyMade™ Solutions
Shipping Condition Dry Ice
Synonym Coxistal, Coxistac
IUPAC Name sodium;(2R)-2-[(2R,5S,6R)-6-[(2S,3S,4S,6R)-6-[(3S,5S,7R,9S,10S,12R,15R)-3-[(2R,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.57.35]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methyloxan-2-yl]butanoate
Molecular Weight (g/mol) 772.98 g/mol
Formula Weight 772.98 amu
Percent Purity >90%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Salinomycin Monosodium Salt ≥0.1 to <1%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.