Learn More
Salinomycin monosodium salt hydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Salinomycin monosodium salt hydrate |
| CAS | 55721-31-8 |
| CAS Max % | 1 |
| Beilstein | 4901827 |
| Synonym | (2R)-2-[(2R,5S,6R)-6-[(3S,5S,7R,9S,10S,12R,15R)-3-[(2R,5R,6S)-5-Ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.5.3]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-ox |
| Molecular Weight (g/mol) | 772.98 g/mol |
| Quantity | 100 mg |