Learn More
Salinomycin, MedChemExpress
Product Code. 16462747
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16462747
Brand: Medchem Express HY15597
MedChemExpress Salinomycin (Procoxacin), a polyether potassium ionophore antibiotic, selectively inhibits the growth of gram-positive bacteria. Salinomycin is a potent inhibitor of Wnt/β-catenin signaling, blocks Wnt-induced LRP6 phosphorylation. Salinomycin (Procoxacin) shows selective activity against human cancer stem cells.
Salinomycin (Procoxacin), a polyether potassium ionophore antibiotic, selectively inhibits the growth of gram-positive bacteria. Salinomycin is a potent inhibitor of Wnt/β-catenin signaling, blocks Wnt-induced LRP6 phosphorylation. Salinomycin (Procoxacin) shows selective activity against human cancer stem cells.
Specifications
Chemical Name or Material Salinomycin
Physical Form Solid
CAS 53003-10-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C42H70O11
For Use With (Application) Cancer-Kinase/protease
Synonym Procoxacin
Solubility Information DMSO : ≥ 36.7 mg/mL (48.87 mM)
SMILES [H][C@]1([C@](C)(CC2)O[C@]32[C@H](O)C=C[C@]4(O[C@]([H])([C@@H](CC)C([C@@H](C)[C@@H](O)[C@H](C)[C@]5([H])O[C@]([C@@H](CC)C(O)=O)([H])CC[C@@H]5C)=O)[C@@H](C)C[C@H]4C)O3)CC[C@@](CC)(O)[C@H](C)O1
Molecular Weight (g/mol) 751.0
Formula Weight 751.0
Percent Purity 95.0%
Grade Research