missing translation for 'onlineSavingsMsg'
Learn More

Salinomycin, MedChemExpress

Product Code. 16426668
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426668

Brand: Medchem Express HY15597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salinomycin (Procoxacin), a polyether potassium ionophore antibiotic, selectively inhibits the growth of gram-positive bacteria. Salinomycin is a potent inhibitor of Wnt/β-catenin signaling, blocks Wnt-induced LRP6 phosphorylation. Salinomycin (Procoxacin) shows selective activity against human cancer stem cells.

Salinomycin (Procoxacin), a polyether potassium ionophore antibiotic, selectively inhibits the growth of gram-positive bacteria. Salinomycin is a potent inhibitor of Wnt/β-catenin signaling, blocks Wnt-induced LRP6 phosphorylation. Salinomycin (Procoxacin) shows selective activity against human cancer stem cells.

Specifications

Chemical Name or Material Salinomycin
Physical Form Solid
CAS 53003-10-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C42H70O11
For Use With (Application) Cancer-Kinase/protease
Synonym Procoxacin
Solubility Information DMSO : ≥ 36.7 mg/mL (48.87 mM)
SMILES [H][C@]1([C@](C)(CC2)O[C@]32[C@H](O)C=C[C@]4(O[C@]([H])([C@@H](CC)C([C@@H](C)[C@@H](O)[C@H](C)[C@]5([H])O[C@]([C@@H](CC)C(O)=O)([H])CC[C@@H]5C)=O)[C@@H](C)C[C@H]4C)O3)CC[C@@](CC)(O)[C@H](C)O1
Molecular Weight (g/mol) 751.0
Formula Weight 751.0
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.