missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salidroside, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Salidroside is a prolyl endopeptidase inhibitor. Salidroside alleviates cachexia symptoms in mouse models of cancer cachexia via activating mTOR signalling. Salidroside protects dopaminergic neurons by enhancing PINK1/Parkin-mediated mitophagy.
Specifications
Specifications
| Chemical Name or Material | Salidroside |
| CAS | 10338-51-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H20O7 |
| Quantity | 5 mg |
| Synonym | Rhodioloside |
| Solubility Information | H2O : ≥ 100 mg/mL (333.00 mM) |
| SMILES | O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OCCC2=CC=C(O)C=C2 |
| Molecular Weight (g/mol) | 300.3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction