missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol Impurity L Acetate Salt, TRC

Product Code. 30417317
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417317 1 mg 1mg
30413065 5 mg 5mg
30412189 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417317 Supplier Toronto Research Chemicals Supplier No. TRCS0855651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salbutamol Impurity L Acetate Salt
Molecular Formula C13 H20 Cl N O3 . C2 H4 O2
Quantity 1 mg
Synonym 5-Chloro-α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-1,3-benzenedimethanol Acetate Salt
InChI Formula InChI=1S/C13H20ClNO3.C2H4O2/c1-13(2,3)15-6-11(17)8-4-9(7-16)12(18)10(14)5-8;1-2(3)4/h4-5,11,15-18H,6-7H2,1-3H3;1H3,(H,3,4)
SMILES CC(=O)O.CC(C)(C)NCC(O)c1cc(Cl)c(O)c(CO)c1
IUPAC Name acetic acid;4-[2-(tert-butylamino)-1-hydroxyethyl]-2-chloro-6-(hydroxymethyl)phenol
Molecular Weight (g/mol) 333.8078
Formula Weight 333.1343
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.