Learn More
Saint-Gobain Norprene™ Pressure Tubing
Click to view available options
Diameter (English) Inner:
1/2 in.
3/4 in.
5/8 in.
Unit Size:
15m
Specifications
Specifications
| Diameter (English) Inner | 1/2 in. |
| Material | Norprene |
| Diameter (English) Outer | 3/4 in. |
| Overall Length (English) | 50 ft. |
| Maximum Pressure | 100 psi |