Learn More
Saint-Gobain Norprene™ Chemical Tubing
Click to view available options
Diameter (English) Inner:
0.13 in.
Unit Size:
15m
Specifications
Specifications
| Diameter (English) Inner | 0.13 in. |
| Material | Norprene |
| Diameter (English) Outer | 1/4 in. |
| Overall Length (English) | 50 ft. |
| Thickness (English) | 0.0625 in. |
| Maximum Pressure | 33 psi |