Learn More
Saint Gobain Life Sciences Chemware™ PTFE-Coated Forceps
Product Code. 12361369
Click to view available options
Length (Metric):
114 mm
Unit Size:
Each
Product Code. 12361369
Brand: Saint Gobain Life Sciences D1069288
Only null left
Add to basket
Add to basket
Chemware™ PTFE-Coated Forceps for handling hot or chemically active materials.
  • Nickel-plated steel covered with an extra heavy-duty coating of PTFE
  • Nonstick surface is inert
  • Coating prevents scratching glass and other delicate surfaces
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Straight
Length (English) 4.5 in.
Material Chrome-plated Steel
Length (Metric) 114 mm
Product Type Forceps
Quantity (No. per Case) 1 Ea.