missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Safranin O, 95%, pure, high purity biological stain

Product Code. p-3776803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10386640 25 g Glass bottle 25g
10628254 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386640 Supplier Thermo Scientific Chemicals Supplier No. 146640250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation. | CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Synonym Basic Red 2, C.I. 50240, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride
PubChem CID 2723800
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

Dye Content 94% min.
Assay Percent Range 95%
Infrared Spectrum Authentic
Formula Weight 350.84
Loss on Drying 10% max.
Grade Pure
Packaging Glass bottle
Color Brown or Brown/Red to Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Safranine O, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Safranine O
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.