missing translation for 'onlineSavingsMsg'
Learn More

SAFit1, MedChemExpress

Product Code. 16456447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456447 5 mg 5mg
16463708 10 mg 10mg
16462836 25 mg 25mg
16430097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16456447 Supplier Medchem Express Supplier No. HY102079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAFit1 is a FK506 binding protein 51 (FKBP51)-specific inhibitor with a Ki of 4±0.3 nM.

SAFit1 is a FK506 binding protein 51 (FKBP51)-specific inhibitor with a Ki of 4±0.3 nM.

Specifications

Chemical Name or Material SAFit1
CAS 1643125-32-9
Color White
Purity Grade Notes Research
Molecular Formula C42H53NO11
Quantity 5 mg
Solubility Information DMSO : ≥ 300 mg/mL (401.14 mM)
SMILES O=C(N1[C@@H](CCCC1)C(O[C@@H](C2=CC(OCC(O)=O)=CC=C2)CCC3=CC(OC)=C(OC)C=C3)=O)[C@@H](C4CCCCC4)C5=CC(OC)=C(OC)C(OC)=C5
Molecular Weight (g/mol) 747.87
Formula Weight 747.87
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.