Learn More
SymLine™ Safety Waste Caps
Product Code. 15960386
Click to view available options
Thread Size:
GL45
S55
S60/61
S90
Includes:
1 GL25 (m), Filling Lance, Connector for Exhaust Filter
1 NPT 3/8 in. (f), Connection for Exhaust Filter
2 PFA-fittings, Tube Connector
2 PFA-fittings, Tube Connector, Collector for HPLC Waste
3 PFA-fittings, Collector for HPLC Waste
5 PFA-fittings, Collector for HPLC Waste
Level Control w/ Shut-off, Connector for Exhaust Filter
Level Control, Cable Connection, Connector for Exhaust Filter
Level Control, Connector for Exhaust Filter
Level Control, Filling Lance, Connector for Exhaust Filter
Unit Size:
1 sheet
Each
Product Code. 15960386
Brand: SymLine™ 106478
Only null left
Add to basket
Add to basket
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
  • Available with different thread sizes for attachment to pipe system
  • Different numbers of hose connections for disposal of liquid waste
  • Available with electronic level sensors that indicate the degree of filling of the disposal container
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifica
For Use With (Application) Waste Disposal
For Use With (Equipment) SymLine System
Includes Level Control, Filling Lance, Connector for Exhaust Filter
Thread Size S55
Type Safety Waste Cap
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato