Learn More
SymLine™ Signalbox and Switch Amplifier Accessories
Accessories for the Built-in Signalbox and the Switch Amplifier.
33.30 EUR - 320.00 EUR
Specifications
For Use With (Application) | Accessorizing Signalbox and Switch Amplifier |
---|
Product Code | Brand | For Use With (Equipment) | Includes | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Product Type | Price | Quantity & Availability | |||
10669290
|
SymLine™
108050 |
Electronic Level Control | Signal Cable (3 m Long) | Signal Cable |
55.40 EUR
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
10104574
|
SymLine™
108037 |
Electronic Level Control | Signal Cable for Electronic Level Control (5 m Long) | Signal Cable |
59.10 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
10202570
|
SymLine™
108038 |
Electronic Level Control | Signal Cable (10 m Long) | Signal Cable |
61.55 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10810571
|
SymLine™
108045 |
Safety Waste Caps | Disc Sensor, Empty Level | Disc Sensor |
320.00 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10337759
|
SymLine™
108048 |
Safety Waste Caps | Disc Sensor, Alarm at Full State | Disc Sensor |
320.00 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
15912531
|
SymLine™
108219 |
Switch Amplifier and Built-in SignalBox | Cable Set | Signal Cable |
95.90 EUR
1 set |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10657145
|
SymLine™
108039 |
Relay Contact, Switch-off Peripheric Instruments | Cable for Relay Contact | Signal Cable |
45.60 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10367589
|
SymLine™
108051 |
Relay Contact | EU Power Supply (220/230 V) | Power Supply |
88.40 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
16593532
|
SymLine™
106590 |
Isolating Switch Amplifier 108278 | Housing for Isolating Switch Amplifier | Housing |
214.00 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10810021
|
SymLine™
900107 |
Level Control Disc Sensors | 10 Dual Lock, Reclosable Fastening Systems for Disc Sensor | Dual Lock Closure |
37.00 EUR
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10121310
|
SymLine™
900108 |
Disc Sensors | Hook and Loop Fastener Tape | Dual Lock Closure |
33.30 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
Description
- LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height
- When container is changed, the alert can be muted via the new touchpad
- Pumps and other external devices can be conveniently triggered via the integrated interface
- According to ATEX Directives, Ex-sensors have to be installed in conjunction with a switch amplifier when used in explosion-prone areas (Ex-zones)
- Signalboxes and switch amplifier must be placed outside of the Ex-zone
- Monitor two filling levels at once
- Touchpad
- Level control through status LEDs
- Optical and acoustic alarm signals
- Compatible with Switch AmplifierATEX: A widely used synonym for the ATEX directives of the European Union. The term ATEX is derived from the French abbreviation for ATmosphere EXplosive. Currently, this directive comprises two EU Directives on explosion prevention, namely the ATEX Equipment Directive 2014/34/EU and the ATEX Workplace Directive 1999/92/EC.
Specifications
Accessorizing Signalbox and Switch Amplifier |