Learn More
SCAT HPLC Safety Caps
Shop All SCAT Europe Products
Click to view available options
Includes:
1 PFA Fitting (3.2 mm OD), Air Valve
2 PFA Fittings (3.2 mm OD), Air Valve
3 PFA Fittings (1.6/2.3/3.2 mm OD), 2 PFA Blind Plugs, Air Valve
3 PFA Fittings (3.2 mm OD), Air Valve
4 PFA Fittings (3.2 mm OD), Air Valve
Unit Size:
Each
Description
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every SafetyCap
Specifications
Specifications
| For Use With (Equipment) | Ground Neck Bottles 29/32 mm |
| Includes | 2 PFA Fittings (3.2 mm OD), Air Valve |
| Material | PTFE |
| Thread Size (Metric) | NS29/32 |
| For Use With (Application) | HPLC Systems |
| Type | Safety Cap II |
| Autoclavable | Autoclavable |