Description
- 3-level coding ensures safe sample and vessel identification.
- Codes on three sides of plates allow flexible reading.
- ECC 200 error correction enables safe code reading even with up to 25% damaged codes.
- Additionally, ID-specific information is available via Eppendorf DataPort for documentation.
- Direct import into eLabInventory and eLabJournal™ software for convenient handling.
- Import to other database systems is possible.
- Manufactured from high-quality polypropylene for the highest sample purity.
Specifications
Specifications
| Barcode | Includes Barcode |
| Format | SBS Standard |
| Array | 16 x 24 |
| Product Line | Eppendorf |
| Color | White, Clear Bottom |
| Product Type | PCR Plate |
| Material | Polypropylene |
| Volume (Metric) Well | 140 μL |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Show More |