Learn More
BD Sabouraud Agar with Chloramphenicol and Cycloheximide
Product Code. 12686147
Click to view available options
:
Sabouraud Agar; Quantity: 20 Plates
Product Code. 12686147
Brand: BD 255504
BD Sabouraud Agar with Chloramphenicol and Cycloheximide is a general purpose medium devised by Sabouraud for the cultivation of dermatophytes.
  • BD Sabouraud Agar with Chloramphenicol and Cycloheximide is a general purpose medium devised for the cultivation of dermatophytes.
  • The low pH of approximately 5.6 and the high glucose concentration is favorable for the growth of all fungi.
TRUSTED_SUSTAINABILITY
Specifications
Media Type Sabouraud Agar
Formulation Pancreatic Digest of Casein 5.0 g/L, Peptic Digest of Animal Tissue g/L, Glucose 40.0 g/L, Chloramphenicol 0.05 g/L, Cycloheximide 0.4 g/L, Agar 23.5 g/L