Learn More
BD Sabouraud Agar with Chloramphenicol and Cycloheximide
Click to view available options
:
Sabouraud Agar; Quantity: 20 Plates
Description
- BD Sabouraud Agar with Chloramphenicol and Cycloheximide is a general purpose medium devised for the cultivation of dermatophytes.
- The low pH of approximately 5.6 and the high glucose concentration is favorable for the growth of all fungi.
Specifications
Specifications
| Media Type | Sabouraud Agar |
| Formulation | Pancreatic Digest of Casein 5.0 g/L, Peptic Digest of Animal Tissue g/L, Glucose 40.0 g/L, Chloramphenicol 0.05 g/L, Cycloheximide 0.4 g/L, Agar 23.5 g/L |