missing translation for 'onlineSavingsMsg'
Learn More

Elma™ S2 Cleaner

Product Code. 12692613
Click to view available options
Volume (Metric):
2.5 L
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 12692613

Brand: Elma™ 8000172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultrasonic cleaning concentrate for industry, laboratory and workshop.

  • Elma tec clean S2 is a liquid, acidic cleaning concentrate for industry, laboratory and workshop.
  • Suitable for stainless steel, precious metals, glass, ceramics and selected plastic.
  • Removes polishing media, lapping paste, mineral grease and oil, rust and lime.
  • It derusts iron metal surfaces.
  • Good for pickling and decalcifying.
  • Check plastics and other metals for resistance against corrosion and tarnish before use.
  • Attacks the surface of light metals (AI, Mg and their alloys).

Specifications

Form Liquid
pH <1
Composition Non-ionic surfactants <5%, Phosphoric acid >30%
Concentration Ultrasonic bath 1 to 5% and for without ultrasonics >3% with tap water
Container Type Can
Content And Storage -5°C to 30°C
Volume (Metric) 2.5 L
compliance-icons
Product Identifier
  • elma tec clean S2
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : phosphoric acid, isotridecanol, ethoxylated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.