missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P5/EDG-8 Rabbit anti-Mouse, Clone: 1196A, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9084-SP
This item is not returnable.
View return policy
Description
S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| S1P5/EDG-8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 53637 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 1196A | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
| Q91X56 | |
| S1pr5 | |
| Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56 | |
| 25 μg | |
| Primary | |
| Detects mouse S1P 5EDG-8 in Western blots. | |
| Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?