Learn More
Veolia Water Systems™ S15 Reverse Osmosis System
Click to view available options
:
Flow Rate: 15L/hour
Description
- Applications range from glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets.
- Features reverse-osmosis-feed-optimized resin mixes to increase the ion exchange capacity of consumables and minimize running costs.
- Quick and easy sanitization and replacement of consumables to reduce maintenance time.
- Designed to be easy to access, whether wall- or bench-mounted, with a convenient dispense tap.
- Can be used with our wrap-around reservoirs to save space whilst optimizing purity.
- Option-S Series systems use multiple purification technologies to produce general laboratory grade water at better than single distilled quality.
- Purification technologies include pre-treatment, reverse osmosis and ion exchange.
Specifications
Specifications
| Capacity | 360 L/24-hour day |
| Flow Rate | 15 L/hr. |
| Inlet Pressure | Without internal boost pump: 60 to 90 psi |
| Purity | Inorganics @ 25°C: 1 to <10MΩ-cm, Total orgnic carbon (TOC): <30ppb |
| Dimensions (L x W x H) | 270 x 410 x 460 mm |
| Weight (Metric) | 14.5 kg without internal boost pump |
| Electrical Requirements | 100-240VAC, 50-60Hz |