missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Rat anti-Mouse, Clone: 493804, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5167-SP
This item is not returnable.
View return policy
Description
S100A11 Monoclonal specifically detects S100A11 in Mouse samples. It is validated for Western Blot.Specifications
| S100A11 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P50543 | |
| S100A11 | |
| E. coli-derived recombinant mouse S100A11 Met1-Ile98 Accession # P50543 | |
| 25 μg | |
| Primary | |
| Detects mouse S100A11 in direct ELISAs and Western blots. In direct ELISAs, ;no cross-reactivity with recombinant mouse S100A1, 4, 6, 8, 9, 10, 13, or 16 is observed, and approximately 75% cross-reactivity with recombinant human (rh) S100A11 is observed. In Western blots, no cross-reactivity with rhS100A11 is observed | |
| Mouse | |
| Purified |
| Western Blot | |
| 493804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6282 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?