missing translation for 'onlineSavingsMsg'
Learn More

(S,R,S)-AHPC-C4-NH2 hydrochloride, MedChemExpress

Product Code. p-200097691
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470146 2 g 2g
16451616 500 mg 500mg
16441209 1 g 1g
16435596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470146 Supplier Medchem Express Supplier No. HY114176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.

(S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.

Specifications

Chemical Name or Material (S,R,S)-AHPC-C4-NH2 hydrochloride
CAS 2245697-83-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H40ClN5O4S
Quantity 2 g
Synonym VH032-C4-NH2 hydrochloride VHL Ligand-Linker Conjugates 13 E3 ligase Ligand-Linker Conjugates 28
Solubility Information DMSO : ≥ 109.6 mg/mL (193.58 mM) ∣H2O : ≥ 50 mg/mL (88.31 mM)
SMILES O=C(N[C@@H](C(C)(C)C)C(N1C[C@H](O)C[C@H]1C(NCC2=CC=C(C=C2)C3=C(C)N=CS3)=O)=O)CCCCN.[H]Cl
Molecular Weight (g/mol) 566.16
Formula Weight 566.16
Percent Purity 95.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.