Learn More
(S)-(-)-Ofloxacin-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Levofloxacin N-methyl-d3 |
| CAS | 2208780-63-4 |
| Molecular Formula | C18 D3 H17 F N3 O4 |
| Quantity | 0.01 g |
| Synonym | (3 S)-9-Fluoro-2,3-dihydro-3-methyl-10-[4-(methyl-d3)-1-piperazinyl]-7-oxo-7 H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, Levofloxacin-D3 (N-methyl-D3), Levofloxacin-d3 |
| InChI Formula | InChI=1 S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10 H,3-6,9H2,1-2H3,(H,24,25)/t10-/m0/s1/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CCN(CC1)c2c(F)cc3C(=O)C(=CN4[C@@H](C)COc2c34)C(=O)O |
| Molecular Weight (g/mol) | 364.386 |
| Formula Weight | 364.163 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |