Learn More
(S)-(+)-Ketoprofen-13C,d3, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-(+)-Ketoprofen-13C,d3 |
| CAS | 1330260-99-5 |
| Molecular Formula | 13C C15 D3 H11 O3 |
| Quantity | 2.5 mg |
| Synonym | (S)-(+)-3-Benzoyl-α-methylbenzeneacetic Acid-13C,d3, (+)-Ketoprofen-13C,d3, (S)-Ketoprofen-13C,d3, (S)-2-(3-Benzoylphenyl)propionic Acid-d3, Dexketoprofen-d3 |
| InChI Formula | InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/t11-/m0/s1/i1+1D3 |
| SMILES | [2H][13C]([2H])([2H])[C@H](C(=O)O)c1cccc(c1)C(=O)c2ccccc2 |
| IUPAC Name | (2S)-2-(3-benzoylphenyl)-3,3,3-trideuterio-(3^{13}C)propanoic acid |
| Molecular Weight (g/mol) | 258.29 |
| Formula Weight | 258.1165 |