Learn More
(S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide, Mikromol™
Product Code. 17347902
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 17347902
Brand: Mikromol MM0236.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
CAS 5439-87-2
Analyte or Component Names (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O5 S
Certifications/Compliance ISO 17025
Synonym (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
InChI Formula InChI=1S/C5H9NO5S/c6-3(5(9)10)1-12(11)2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-,12?/m0/s1
SMILES N[C@@H](CS(=O)CC(=O)O)C(=O)O
IUPAC Name (2R)-2-amino-3-(carboxymethylsulfinyl)propanoic acid
Molecular Weight (g/mol) 195.19
Quantity 100 mg
Formula Weight 195.0201 g/mol