missing translation for 'onlineSavingsMsg'
Learn More

(S)-Bepotastine Besylate, TRC

Product Code. 30345157
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30345157

Brand: Toronto Research Chemicals TRCB3170005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Bepotastine Besylate
CAS 190786-44-8
Molecular Formula C21 H25 Cl N2 O3 . C6 H6 O3 S
Quantity 5 mg
Synonym 1-Piperidinebutanoic acid, 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy]-, benzenesulfonate (1:1), 1-Piperidinebutanoic acid, 4-[(4-chlorophenyl)-2-pyridinylmethoxy]-, (S)-, monobenzenesulfonate, 1-Piperidinebutanoic acid, 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy]-, monobenzenesulfonate (9CI), (S)-4-[4-(4-chlorophenyl-2-pyridylmethoxy)piperidinyl]butyric acid benzenesulfonate, BB, Bepotastine benzenesulfonate, Bepotastine benzenesulfonate salt, Bepotastine besilate, Bepotastine besylate, Bepreve, Betatastine, TAU 284, Talion, Tarion
InChI Formula InChI=1S/C21H25ClN2O3.C6H6O3S/c22-17-8-6-16(7-9-17)21(19-4-1-2-12-23-19)27-18-10-14-24(15-11-18)13-3-5-20(25)26;7-10(8,9)6-4-2-1-3-5-6/h1-2,4,6-9,12,18,21H,3,5,10-11,13-15H2,(H,25,26);1-5H,(H,7,8,9)/t21-;/m0./s1
SMILES OC(=O)CCCN1CCC(CC1)O[C@@H](c2ccc(Cl)cc2)c3ccccn3.OS(=O)(=O)c4ccccc4
IUPAC Name benzenesulfonic acid;4-[4-[(S)-(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoic acid
Molecular Weight (g/mol) 547.06
Formula Weight 546.1591
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.