missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Bepotastine Besylate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | (S)-Bepotastine Besylate |
| CAS | 190786-44-8 |
| Molecular Formula | C21 H25 Cl N2 O3 . C6 H6 O3 S |
| Quantity | 10 mg |
| Synonym | 1-Piperidinebutanoic acid, 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy]-, benzenesulfonate (1:1), 1-Piperidinebutanoic acid, 4-[(4-chlorophenyl)-2-pyridinylmethoxy]-, (S)-, monobenzenesulfonate, 1-Piperidinebutanoic acid, 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy]-, monobenzenesulfonate (9CI), (S)-4-[4-(4-chlorophenyl-2-pyridylmethoxy)piperidinyl]butyric acid benzenesulfonate, BB, Bepotastine benzenesulfonate, Bepotastine benzenesulfonate salt, Bepotastine besilate, Bepotastine besylate, Bepreve, Betatastine, TAU 284, Talion, Tarion |
| InChI Formula | InChI=1S/C21H25ClN2O3.C6H6O3S/c22-17-8-6-16(7-9-17)21(19-4-1-2-12-23-19)27-18-10-14-24(15-11-18)13-3-5-20(25)26;7-10(8,9)6-4-2-1-3-5-6/h1-2,4,6-9,12,18,21H,3,5,10-11,13-15H2,(H,25,26);1-5H,(H,7,8,9)/t21-;/m0./s1 |
| SMILES | OC(=O)CCCN1CCC(CC1)O[C@@H](c2ccc(Cl)cc2)c3ccccn3.OS(=O)(=O)c4ccccc4 |
| IUPAC Name | benzenesulfonic acid;4-[4-[(S)-(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoic acid |
| Molecular Weight (g/mol) | 547.06 |
| Formula Weight | 546.1591 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction