missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Antibody (PDS-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80937
This item is not returnable.
View return policy
Description
S-arrestin Monoclonal specifically detects S-arrestin in Human, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| S-arrestin | |
| Monoclonal | |
| 0.77 mg/mL | |
| Western Blot : 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence : 1:100-1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen : 1:100-1:200 | |
| arrestin 1, RP47,48 kDa protein, S-antigen; retina and pineal gland (arrestin), S-arrestin, visual arrestin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Pig, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| PDS-1 | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| SAG | |
| Porcine retinal S-arrestin [Swiss-Prot# P79260] | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 6295 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction