missing translation for 'onlineSavingsMsg'
Learn More

S-Adenosyl-L-methionine disulfate tosylate, MedChemExpress

Product Code. 16455457
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16455457

Brand: Medchem Express HYW017770

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress S-Adenosyl-L-methionine disulfate tosylate (Ademetionine disulfate tosylate) is the principal biological methyl donor synthesized in all mammalian cells but most abundantly in the liver.

S-Adenosyl-L-methionine disulfate tosylate (Ademetionine disulfate tosylate) is the principal biological methyl donor synthesized in all mammalian cells but most abundantly in the liver.

Specifications

Chemical Name or Material S-Adenosyl-L-methionine disulfate tosylate
CAS 97540-22-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H34N6O16S4
Synonym Ademetionine disulfate tosylateS-Adenosyl methionine disulfate tosylateAdoMet disulfate tosylate
Solubility Information DMSO : ≥ 50 mg/mL (65.21 mM) ∣H2O : 33.33 mg/mL (43.47 mM; Need ultrasonic)
SMILES N[C@@H](CC[S+](C[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O)O)C)C(O)=O.O=S(O)([O-])=O.O=S(O)(O)=O.OS(=O)(C4=CC=C(C)C=C4)=O
Molecular Weight (g/mol) 766.8
Quantity 50 mg
Formula Weight 766.8
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.