Learn More
(S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid, TRC
Product Code. 30417874
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417874 1 g
30417878 500 mg
30417877 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30417874 Supplier Toronto Research Chemicals Supplier No. TRCT2051651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1026987-94-9
Molecular Formula C23H40N2O9
Molecular Weight (g/mol) 488.57
Synonym DUPA(OtBu)-OH
IUPAC Name (4S)-4-[[(2S)-1,5-bis[(2-methylpropan-2-yl)oxy]-1,5-dioxopentan-2-yl]carbamoylamino]-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CC[C@H](NC(=O)N[C@@H](CCC(=O)O)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C23H40N2O9/c1-21(2,3)32-17(28)13-11-15(19(30)34-23(7,8)9)25-20(31)24-14(10-12-16(26)27)18(29)33-22(4,5)6/h14-15H,10-13H2,1-9H3,(H,26,27)(H2,24,25,31)/t14-,15-/m0/s1
Formula Weight 488.2734
Chemical Name or Material (S)-5-Tert-butoxy-4-(3-((s)-1,5-di-tert-butoxy-1,5-dioxopentan-2-yl)ureido)-5-oxopentanoic Acid