missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-2-methyl-4-pentenoic acid, 98%, ee 99+%

Product Code. p-7036604
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401776 250 mg 250mg
15491766 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401776 Supplier Thermo Scientific Alfa Aesar Supplier No. H54922.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96886-55-4 | C6H11NO2 | 129.159 g/mol

It is used in organic synthesis and as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis and as an pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96886-55-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00145248
InChI Key QMBTZYHBJFPEJB-LURJTMIESA-N
Synonym s-2-amino-2-methylpent-4-enoic acid, s-2-amino-2-methyl-4-pentenoic acid, l-alpha-allylalanine, 2s-2-amino-2-methylpent-4-enoic acid, alpha-allyl-l-ala, r-2-amino-2-methyl-4-pentenoic acid, alpha-methyl-l-allylglycine, s---alpha-allylalanine, 4-pentenoic acid, 2-amino-2-methyl-, 2s, s---2-amino-2-methyl-4-pentenoic acid
PubChem CID 2724753
IUPAC Name (2S)-2-amino-2-methylpent-4-enoic acid
SMILES CC(CC=C)(C(=O)O)N

Specifications

Melting Point 285°C
Quantity 250 mg
Solubility Information Very soluble in water.
Optical Rotation −14° (c=1.3 in 1N HCl)
Formula Weight 129.16
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-2-Amino-2-methyl-4-pentenoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.