Learn More
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%
Product Code. 11392297
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392297 1 g 1g
11302307 5 g 5g
15420827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11392297 Supplier Thermo Scientific Alfa Aesar Supplier No. L09855.03
Request Bulk or Custom Format
CAS: 22204-53-1 | C14H14O3 | 230.263 g/mol
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22204-53-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD00010500
InChI Key CMWTZPSULFXXJA-VIFPVBQESA-N
Synonym naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno
PubChem CID 156391
ChEBI CHEBI:7476
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
Specifications
Melting Point 154°C to 156°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 3591067
Merck Index 14,6417
Solubility Information Insoluble in water.
Optical Rotation +66° (c=1 in Chloroform)
Formula Weight 230.26
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid