missing translation for 'onlineSavingsMsg'
Learn More

Rubicon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen Rubicon
Concentration 0.47 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16851035
View Documents
Proteintech
21444-1-AP-20UL
20 μL
145.00 EUR
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16841035
View Documents
Proteintech
21444-1-AP-150UL
150 μL
410.00 EUR
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Two Beclin-1-interacting proteins, the run domain Beclin-1 interacting and cysteine-rich containing protein (Rubicon) and ATG14L, reciprocally regulate autophagy at different stages. Knockdown of Rubicon caused enhancement of autophagy while that of ATG14L caused a defect in autophagosome formation. Rubicon functions as part of a Beclin-1-PIK3C3-containing autophagy complex and is also an essential, positive regulator of the NAPDH oxidase complex. Upon microbial infection or TLR2 activation, Rubicon interacts with the CYBA subunit of the NAPDH oxidase complex, leading to a burst of reactive oxygen species and inflammatory cytokines.
Specifications

Specifications

Rubicon
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
Q80U62, Q92622
100502698, 303885, 9711
Rubicon Fusion Protein Ag13715
Primary
-20°C
RUBCN
0.47 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Baron, KIAA0226, RUBCN, Rubicon
Rubcn
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.