Learn More
Fisherbrand™ Solid Rubber Stoppers
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
5.05 EUR - 37.25 EUR
Specifications
Color | Red |
---|---|
Material | Rubber |
Product Type | Stopper |
For Use With (Application) | Laboratory use |
Product Code | Brand | Diameter (Metric) Bottom | Stopper Number | Height (Metric) | Diameter (Metric) Top | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Stopper Number | Height (Metric) | Diameter (Metric) Top | Price | Quantity & Availability | ||
11512922
|
Fisherbrand™
|
10 mm | 10 | 20mm | 12.5 mm |
6.20 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11532922
|
Fisherbrand™
|
13 mm | 13 | 24mm | 16 mm |
6.80 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11542922
|
Fisherbrand™
|
15 mm | 15 | 24mm | 18 mm |
8.00 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11562922
|
Fisherbrand™
|
19 mm | 19 | 28mm | 22.5 mm |
12.55 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11572922
|
Fisherbrand™
|
21 mm | 21 | 28mm | 24.5 mm |
14.60 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11502932
|
Fisherbrand™
|
27 mm | 27 | 32mm | 31 mm |
13.35 EUR
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11512932
|
Fisherbrand™
|
29 mm | 29 | 32mm | 33 mm |
14.65 EUR
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
10355351
|
Fisherbrand™
|
31 mm | 31 | 35mm | 36 mm |
21.20 EUR
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11532932
|
Fisherbrand™
|
33 mm | 33 | 38.5mm | 38.5 mm |
24.85 EUR
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11552932
|
Fisherbrand™
|
37 mm | 37 | 38.5mm | 42.5 mm |
29.25 EUR
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
11562932
|
Fisherbrand™
|
39 mm | 39 | 38.5mm | 44.5 mm |
33.85 EUR
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||
11542912
|
Fisherbrand™
|
3 mm | 3 | 12mm | 4.5 mm |
8.10 EUR
Pack of 50 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
10558431
|
Fisherbrand™
|
41 mm | 41 | 42mm | 47 mm |
24.50 EUR
Pack of 5 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||
11582932
|
Fisherbrand™
|
43 mm | 43 | 42mm | 49 mm |
23.20 EUR
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11502942
|
Fisherbrand™
|
49 mm | 49 | 42mm | 55 mm |
30.60 EUR
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11592932
|
Fisherbrand™
|
45 mm | 45 | 42mm | 51 mm |
28.25 EUR
Pack of 5 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
11552912
|
Fisherbrand™
|
4 mm | 4 | 14mm | 5.8 mm |
8.20 EUR
Pack of 50 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11512942
|
Fisherbrand™
|
53 mm | 53 | 45.5mm | 59.5 mm |
17.40 EUR
Pack of 2 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11562912
|
Fisherbrand™
|
5 mm | 5 | 16mm | 7 mm |
11.90 EUR
Pack of 50 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||
11557582
|
Fisherbrand™
|
71 mm | 71 | 49mm | 78 mm |
17.80 EUR
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11587582
|
Fisherbrand™
|
86 mm | 86 | 54mm | 96 mm |
29.30 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||
11567582
|
Fisherbrand™
|
75 mm | 75 | 54mm | 85 mm |
17.95 EUR
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11582912
|
Fisherbrand™
|
7 mm | 7 | 16mm | 9 mm |
5.05 EUR
Pack of 20 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11592912
|
Fisherbrand™
|
8 mm | 8 | 20mm | 10.5 mm |
5.65 EUR
Pack of 20 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
11507592
|
Fisherbrand™
|
99 mm | 99 | 54mm | 109 mm |
37.25 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||
10151801
|
Fisherbrand™
|
9 mm | 9 | 20mm | 11.5 mm |
6.05 EUR
Pack of 20 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
10191411
|
Fisherbrand™
|
43 mm | 43 | 42mm | 49 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
- Temperature range -40° to +70°C
- Not suitable for use with petroleum oils and hydrocarbon solvents
Specifications
Red | |
Rubber | |
Stopper | |
Laboratory use |