Learn More
ROBU Rubber Sleeves
Click to view available options
Diameter (Metric) Outer:
26 mm
33 mm
41 mm
Unit Size:
Pack of 5
Description
- Seals filter crucible and corresponding adapter
Specifications
Specifications
| Description | For 8 mL Filter Crucibles |
| Diameter (Metric) Inner | 20 mm |
| Material | Natural Rubber |
| Quantity | 5/Pk. |
| Diameter (Metric) Outer | 26 mm |