Learn More
Saint-Gobain Rubber Pipet Suction Bulb
Click to view available options
:
Capacity: 5mL; Yellow; Color Coding
Description
- For liquid handling
Specifications
Specifications
| Color | Yellow |
| Material | Rubber |
| Type | Suction Bulb |
| Volume (Metric) | 5 mL |
| For Use With (Application) | For fast and easy pipetting |
| Quantity | 1 Each |