Learn More
Saint Gobain Life Sciences Rubber Folding Skirted Stopper
Click to view available options
Color:
Transparent
Unit Size:
Pack of 100
Description
- Two-part folding skirt stoppers
- Designed with a double seal to keep container contents secure
- Ideal for securing glass, metal and plastic containers
- The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
- Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
- For test tubes
- Easy to install
Specifications
Specifications
| Diameter (Metric) | 7.1mm |
| Quantity | 100/ck. |
| Color | Transparent |
| Material | Rubber |
| Product Type | Rubber Stopper |