missing translation for 'onlineSavingsMsg'
Learn More

RSV604, MedChemExpress

Product Code. p-200099059
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498177

Brand: Medchem Express HY12993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RSV604 (A-60444) is an inhibitor of respiratory syncytial virus (RSV) replication. RSV604 targets the nucleocapsid protein, with a Kd of 1.6 μM. RSV604 displays submicromolar activity against numerous clinical isolates of both the A and B subtypes of RSV (average EC50s=0.8 μM).

RSV604 (A-60444) is an inhibitor of respiratory syncytial virus (RSV) replication. RSV604 targets the nucleocapsid protein, with a Kd of 1.6 μM. RSV604 displays submicromolar activity against numerous clinical isolates of both the A and B subtypes of RSV (average EC50s=0.8 μM).

Specifications

Chemical Name or Material RSV604
CAS 676128-63-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H17FN4O2
Quantity 5 mg
Synonym A-60444
Solubility Information DMSO : 100 mg/mL (257.47 mM; Need ultrasonic)
SMILES O=C1[C@@H](NC(NC2=C(F)C=CC=C2)=O)N=C(C3=CC=CC=C3)C4=CC=CC=C4N1
Molecular Weight (g/mol) 388.39
Formula Weight 388.39
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.