missing translation for 'onlineSavingsMsg'
Learn More

RSV604, MedChemExpress

Product Code. 16498177
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498177 5 mg 5mg
16476056 100 mg 100mg
16455468 10 mg 10mg
16431647 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498177 Supplier Medchem Express Supplier No. HY12993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RSV604 (A-60444) is an inhibitor of respiratory syncytial virus (RSV) replication. RSV604 targets the nucleocapsid protein, with a Kd of 1.6 μM. RSV604 displays submicromolar activity against numerous clinical isolates of both the A and B subtypes of RSV (average EC50s=0.8 μM).

RSV604 (A-60444) is an inhibitor of respiratory syncytial virus (RSV) replication. RSV604 targets the nucleocapsid protein, with a Kd of 1.6 μM. RSV604 displays submicromolar activity against numerous clinical isolates of both the A and B subtypes of RSV (average EC50s=0.8 μM).

Specifications

Chemical Name or Material RSV604
CAS 676128-63-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H17FN4O2
Quantity 5 mg
Synonym A-60444
Solubility Information DMSO : 100 mg/mL (257.47 mM; Need ultrasonic)
SMILES O=C1[C@@H](NC(NC2=C(F)C=CC=C2)=O)N=C(C3=CC=CC=C3)C4=CC=CC=C4N1
Molecular Weight (g/mol) 388.39
Formula Weight 388.39
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.