missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSU1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16586471
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16586471 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16586471 Supplier Invitrogen™ Supplier No. PA588161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSKSLKKLVE ESREKNQPEV DMSDRGISNM LDVNGLFTLS HITQLVLSHN KLTMVPPNIA ELKNLEVLNF FNNQIEELPT QISSLQKLKH LNLGMNRLNT LPRGFGSLPA LEVLDLTYNN LSENSLPGNF FYLTTLRALY LSDNDFEILP PDIGKLTKLQ ILSLRDNDLI SLPKEIGELT QLKELHIQGN RLTVLPPELG NLDLTGQKQV FKAENNPWVT PIADQFQLGV SHVFEYIRSE TYKYLYGRHM QANPEPPKKN NDKSKKISRK PLAAKNR; Positive Samples: A-431, U-87MG, B-cell, Skov3, Mouse heart, Mouse lung, Mouse kidney, Rat heart, Rat lung.

RSU1 is a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the protein was initially isolated based on its ability to inhibit v-Ras transformation. This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSU1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RSU1
Gene Accession No. Q01730, Q15404
Gene Alias cubilin; ras suppressor protein 1; etID12695; FLJ31034; LOC553276 protein; ras suppressor protein 1; ras suppressor protein 1 isoform 1; ras suppressor protein 1 variant 1; ras suppressor protein 1 variant 2; ras suppressor protein 1 variant 3; ras suppressor protein 1 variant 5; Ras suppressor protein 1-like protein; RSP1; RSP-1; rsu1; Rsu-1; RsuI; si:dz63m2.1; Unknown (protein for MGC:126965)
Gene Symbols RSU1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human RSU1 (NP_0365571).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20163, 6251, 680419
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.