missing translation for 'onlineSavingsMsg'
Learn More

RSL3, MedChemExpress

Product Code. p-200099790
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492848

Brand: Medchem Express HY100218A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells.

RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells.

Specifications

Chemical Name or Material RSL3
CAS 1219810-16-8
Color White
Purity Grade Notes Research
Molecular Formula C23H21ClN2O5
Quantity 10 mg
Synonym (1S,3R)-RSL3
Solubility Information DMSO : 250 mg/mL (567.05 mM; Need ultrasonic)
SMILES O=C([C@H]1CC2=C([C@H](C3=CC=C(C(OC)=O)C=C3)N1C(CCl)=O)NC4=C2C=CC=C4)OC
Molecular Weight (g/mol) 440.88
Formula Weight 440.88
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.