Learn More
Tocris Bioscience™ (RS)-3,5-DHPG
GREENER_CHOICE
Selective group I mGlu agonist
Brand:  Tocris Bioscience™ 0342/50
Encompass
Product Code. 13427573
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
(RS)-3,5-DHPG is a selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.
S-enantiomer also available.
Specifications
Specifications
(RS)-3,5-Dihydroxyphenylglycine
>99%
Soluble to 10mM in water
19641-83-9
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions