missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KL1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 470.00 EUR
Specifications
| Antigen | RPS6KL1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331766
|
Novus Biologicals
NBP3-04434-20UL |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347884
|
Novus Biologicals
NBP3-04434-100UL |
100 μL |
470.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPS6KL1 Polyclonal specifically detects RPS6KL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RPS6KL1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.11.1, FLJ35734, MGC11287, ribosomal protein S6 kinase-like 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human RPS6KL1 (NP_113652.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 50% glycerol, pH7.3. | |
| 83694 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title