missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS4X Polyclonal Antibody
GREENER_CHOICE

Product Code. 15603885
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15603885 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15603885 Supplier Invitrogen™ Supplier No. PA549949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RPS4X protein.

Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS4X
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPS4X
Gene Accession No. P62701, P62702, P62703
Gene Alias 40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4
Gene Symbols RPS4X
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human RPS4X.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362640, 20102, 6191
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.