missing translation for 'onlineSavingsMsg'
Learn More

RPA/p70 (Replication Protein A) Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Product Code. 12642057 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
Concentration:
1mg/mL
200μg/mL
Unit Size:
100µL
200µL
500µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration unitSize
12642057 500 μL 200μg/mL 500µL
12652057 100 μL 1mg/mL 100µL
12662057 200 μL 1mg/mL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12642057 Supplier Epredia Supplier No. MS692P1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide accurate, reproducible results Epredia™ RPA/p70 (Replication Protein A) Ab-2, Mouse Monoclonal Antibody.

Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase α and δ. RPA70 is composed of three domains: an N-terminal domain that is not required for ssDNA binding or SV40 replication, a central DNA-binding domain, and a C-terminal domain that is essential for subunit interactions.

Host Species: Mouse

Clone: 2H10 or 70C or αSSB70C

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human RPA protein

Molecular Weight: 70kDa

Positive Control: HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA/p70 (Replication Protein A) Ab-2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H10 or 70C or alphaSSB70C
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human RPA protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.