Learn More
Kartell Round Bowl
Click to view available options
Capacity (Metric):
10 L
Unit Size:
Pack of 10
Description
- Sturdy structure
- Two grips have been designed in the lower part of the rim to help carriage
Specifications
Specifications
| Autoclavable | Autoclavable |
| Bottom Shape | Flat |
| Capacity (Metric) | 10 L |
| Color | White |
| Disposable | Reusable |
| Material | Polypropylene |
| Shape | Round |
| Sterility | Non-sterile |
| Type | Dish |
| Volume (Metric) Working | 10 L |
| Show More |